1-Aminocyclopropancarbonsäure
Lieferant: FLUOROCHEM
Synonyme:
1-Amino-1-cyclopropanecarboxylic acid
043411-5G
FLUO043411-5GEA
79.1
EUR
FLUO043411-5G
1-Aminocyclopropancarbonsäure
1-Aminocyclopropanecarboxylic acid
Formel:
C₄H₇NO₂ Molecular Weight: 101,11 g/mol |
MDL:
MFCD00009944 CAS-Nummer: 22059-21-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...