4-Brom-2-methylbenzotrifluorid

Lieferant: FLUOROCHEM

Synonyme: 4-Bromo-2-methyl-1-(trifluoromethyl)benzene

044187-1G
FLUO044187-1GEA 55.4 EUR
FLUO044187-1G
4-Brom-2-methylbenzotrifluorid
4-Brom-2-methylbenzotrifluorid
Formel: C₈H₆BrF₃
Molecular Weight: 239,0325 g/mol
MDL: MFCD11100526
CAS-Nummer: 936092-88-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR