4,4-Difluorcyclohexanon
Lieferant: FLUOROCHEM
044463-5G
FLUO044463-5GEA
136
EUR
FLUO044463-5G
4,4-Difluorcyclohexanon
4,4-Difluorocyclohexanone
Formel:
C₆H₈F₂O Molecular Weight: 134,12 g/mol |
MDL:
MFCD08146635 CAS-Nummer: 22515-18-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...