2-Fluor-5-nitroanisol
Lieferant: FLUOROCHEM
Synonyme:
4-Fluor-3-methoxynitrobenzol
045066-25G
FLUO045066-25GEA
200
EUR
FLUO045066-25G
2-Fluor-5-nitroanisol
2-Fluor-5-nitroanisol
Formel:
C₇H₆FNO₃ Molecular Weight: 171,13 g/mol |
MDL:
MFCD07782081 CAS-Nummer: 454-16-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...