3-Brom-5-chlor-2-fluorpyridin

Lieferant: FLUOROCHEM

045837-1G
FLUO045837-1GEA 32.1 EUR
FLUO045837-1G
3-Brom-5-chlor-2-fluorpyridin
Formel: C₅H₂BrClFN
MDL: MFCD04972409
CAS-Nummer: 884494-87-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR