2,4-Difluor-N-methylanilin
Lieferant: FLUOROCHEM
045886-1G
FLUO045886-1GEA
0
EUR
FLUO045886-1G
2,4-Difluor-N-methylanilin
2,4-Difluor-N-methylanilin
Formel:
C₇H₇F₂N Molecular Weight: 143,14 g/mol Siedepunkt: 94…96 °C (15 mmHg) |
MDL:
MFCD00042472 CAS-Nummer: 138564-16-6 UN: 2810 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...