2-Fluor-6-nitrobenzoesäure

Lieferant: FLUOROCHEM

Synonyme: 6-Fluoro-2-nitrobenzoic acid

046128-5G
FLUO046128-5GEA 48.9 EUR
FLUO046128-5G
2-Fluor-6-nitrobenzoesäure
2-Fluoro-6-nitrobenzoic acid
Formel: C₇H₄FNO₄
Molecular Weight: 185,11 g/mol
MDL: MFCD01862079
CAS-Nummer: 385-02-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR