Bis(tri-tert-butylphosphin)palladium(0)

Lieferant: FLUOROCHEM

Synonyme: Bis(tri-tert-butylphosphine)palladium, Bis(tri-t-butylphosphine)palladium(0)

046133-1G
FLUO046133-1GEA 294 EUR
FLUO046133-1G
Bis(tri-tert-butylphosphin)palladium(0)
Bis(tri-tert-butylphosphin)palladium(0)
Formel: C₂₄H₅₄P₂Pd
Molecular Weight: 511.05 g/mol
MDL: MFCD03094580
CAS-Nummer: 53199-31-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR