3,5-Difluorpicolinsäure

Lieferant: FLUOROCHEM

Synonyme: 3,5-Difluoropyridine-2-carboxylic acid

046203-5G
FLUO046203-5GEA 107 EUR
FLUO046203-5G
3,5-Difluorpicolinsäure
3,5-Difluorpicolinsäure
Formel: C₆H₃F₂NO₂
Molecular Weight: 159,09 g/mol
MDL: MFCD05664124
CAS-Nummer: 745784-04-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR