2-Bromcarbazol

Lieferant: FLUOROCHEM

Synonyme: 2-Bromo-9H-carbazole

046298-1G
FLUO046298-1GEA 121 EUR
FLUO046298-1G
2-Bromcarbazol
2-Bromocarbazole
Formel: C₁₂H₈BrN
Molecular Weight: 246,11 g/mol
MDL: MFCD09750430
CAS-Nummer: 3652-90-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR