2,4-Dichlorchinazolin

Lieferant: FLUOROCHEM

047662-5G
FLUO047662-5GEA 0 EUR
FLUO047662-5G
2,4-Dichlorchinazolin
2,4-Dichloroquinazoline
Formel: C₈H₄Cl₂N₂
Molecular Weight: 199,04 g/mol
MDL: MFCD00091950
CAS-Nummer: 607-68-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR