4-Brombenzamidin Hydrochlorid

Lieferant: FLUOROCHEM

Synonyme: 4-Brombenzoesäureamidin Hydrochlorid, 4-Bromphenylamidin Hydrochlorid

048724-5G
FLUO048724-5GEA 107 EUR
FLUO048724-5G
4-Brombenzamidin Hydrochlorid
4-Brombenzamidin Hydrochlorid
Formel: C₇H₈BrClN₂
Molecular Weight: 235,51 g/mol
MDL: MFCD00464967
CAS-Nummer: 55368-42-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR