5-Fluor-2-pyridincarbonitril

Lieferant: FLUOROCHEM

Synonyme: 2-Cyan-5-fluorpyridin, 5-Fluor-2-pyridincarbonsäurenitril

049469-5G
FLUO049469-5GEA 87.9 EUR
FLUO049469-5G
5-Fluor-2-pyridincarbonitril
5-Fluor-2-pyridincarbonitril
Formel: C₆H₃FN₂
Molecular Weight: 122,1 g/mol
MDL: MFCD08741371
CAS-Nummer: 327056-62-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR