4-Hydroxy-3-iodbenzotrifluorid

Lieferant: FLUOROCHEM

Synonyme: 2-Iodo-4-(trifluoromethyl)phenol

049768-1G
FLUO049768-1GEA 125 EUR
FLUO049768-1G
4-Hydroxy-3-iodbenzotrifluorid
4-Hydroxy-3-iodbenzotrifluorid
Formel: C₇H₄F₃IO
MDL: MFCD12922960
CAS-Nummer: 463976-21-8
UN: 3264
ADR: 8,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR