2-Formyl-5-methoxypyridin

Lieferant: FLUOROCHEM

Synonyme: 5-Methoxypyridine-2-carbaldehyde, 5-Methoxypicolinaldehyde

050083-1G
FLUO050083-1GEA 377 EUR
FLUO050083-1G
2-Formyl-5-methoxypyridin
2-Formyl-5-methoxypyridin
Formel: C₇H₇NO₂
Molecular Weight: 137,1360 g/mol
MDL: MFCD08234867
CAS-Nummer: 22187-96-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR