2-Amino-5-fluor-4-methylpyridin

Lieferant: FLUOROCHEM

050234-5G
FLUO050234-5GEA 260 EUR
FLUO050234-5G
2-Amino-5-fluor-4-methylpyridin
2-Amino-5-fluor-4-methylpyridin
Formel: C₆H₇FN₂
Molecular Weight: 126,13 g/mol
MDL: MFCD06659479
CAS-Nummer: 301222-66-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR