1-(2-Tetrahydropyranyl)-1H-pyrazol-5-carbaldehyd
Lieferant: FLUOROCHEM
Synonyme:
1-(Oxan-2-yl)pyrazole-5-carbaldehyde
060909-1G
FLUO060909-1GEA
306
EUR
FLUO060909-1G
1-(2-Tetrahydropyranyl)-1H-pyrazol-5-carbaldehyd
1-(2-Tetrahydropyranyl)-1H-pyrazol-5-carbaldehyd
Formel:
C₉H₁₂N₂O₂ |
MDL:
MFCD07186538 CAS-Nummer: 957483-88-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...