3-Amino-6-methylpyridazin

Lieferant: FLUOROCHEM

064948-1G
FLUO064948-1GEA 61.8 EUR
FLUO064948-1G
3-Amino-6-methylpyridazin
3-Amino-6-methylpyridazin
Formel: C₅H₇N₃
Molecular Weight: 109,13 g/mol
MDL: MFCD01569899
CAS-Nummer: 18591-82-7
EINECS: 242-430-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR