4-Brom-5-chloranthranilsäure

Lieferant: FLUOROCHEM

Synonyme: 2-Amino-4-bromo-5-chlorobenzoic acid

077797-1G
FLUO077797-1GEA 75.6 EUR
FLUO077797-1G
4-Brom-5-chloranthranilsäure
4-Brom-5-chloranthranilsäure
Formel: C₇H₅BrClNO₂
Molecular Weight: 250,4800 g/mol
MDL: MFCD10000554
CAS-Nummer: 150812-32-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR