3-Brompyridazin
Lieferant: FLUOROCHEM
078228-1G
FLUO078228-1GEA
194
EUR
FLUO078228-1G
3-Brompyridazin
3-Bromopyridazine
Formel:
C₄H₃BrN₂ Molecular Weight: 158.9840 g/mol |
MDL:
MFCD10688587 CAS-Nummer: 88491-61-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...