5-Amino-2-chlorbenzonitril

Lieferant: FLUOROCHEM

Synonyme: 4-Chloro-3-cyanoaniline

078416-5G
FLUO078416-5GEA 92.9 EUR
FLUO078416-5G
5-Amino-2-chlorbenzonitril
5-Amino-2-chlorbenzonitril
Formel: C₇H₅ClN₂
Molecular Weight: 152,5800 g/mol
MDL: MFCD00800459
CAS-Nummer: 35747-58-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR