Bis(2,4-dimethoxybenzyl)amin

Lieferant: FLUOROCHEM

079530-5G
FLUO079530-5GEA 117 EUR
FLUO079530-5G
Bis(2,4-dimethoxybenzyl)amin
Bis(2,4-dimethoxybenzyl)amin
Formel: C₁₈H₂₃NO₄
Molecular Weight: 317,3800 g/mol
MDL: MFCD00040480
CAS-Nummer: 20781-23-1
EINECS: 244-037-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR