2-Brom-6-iodbenzonitril

Lieferant: FLUOROCHEM

079665-1G
FLUO079665-1GEA 117 EUR
FLUO079665-1G
2-Brom-6-iodbenzonitril
2-Brom-6-iodbenzonitril
Formel: C₇H₃BrIN
Molecular Weight: 307,9139 g/mol
MDL: MFCD16619725
CAS-Nummer: 1245648-93-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR