2-(1-Hydroxyethyl)pyridin

Lieferant: FLUOROCHEM

Synonyme: 1-(Pyridin-2-yl)ethanol

088389-1G
FLUO088389-1GEA 36.9 EUR
FLUO088389-1G
2-(1-Hydroxyethyl)pyridin
2-(1-Hydroxyethyl)pyridine
Formel: C₇H₉NO
Molecular Weight: 123,1525 g/mol
MDL: MFCD06245419
CAS-Nummer: 18728-61-5
EINECS: 242-539-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR