2,2'-Dipyridyldisulfid
Lieferant: FLUOROCHEM
Synonyme:
2,2'-Dithiodipyridin
136900-25G
FLUO136900-25GEA
106
EUR
FLUO136900-25G
2,2'-Dipyridyldisulfid
2,2'-Dipyridyldisulfid
Formel:
C₁₀H₈N₂S₂ Molecular Weight: 220,32 g/mol Siedepunkt: 348 °C (1013 hPa) Schmelzpunkt: 57…61 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00006287 CAS-Nummer: 2127-03-9 EINECS: 218-343-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...