3-Iodbenzonitril
Lieferant: FLUOROCHEM
147901-5G
FLUO147901-5GEA
45.9
EUR
FLUO147901-5G
3-Iodbenzonitril
3-Iodbenzonitril
Formel:
C₇H₄IN Molecular Weight: 229,02 g/mol |
MDL:
MFCD00079762 CAS-Nummer: 69113-59-3 UN: 3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...