6-Iod-m-toluylsäure

Lieferant: FLUOROCHEM

148705-25G
FLUO148705-25GEA 94.1 EUR
FLUO148705-25G
6-Iod-m-toluylsäure
6-Iod-m-toluylsäure
Formel: C₈H₇IO₂
Molecular Weight: 262,05 g/mol
MDL: MFCD00079765
CAS-Nummer: 52548-14-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR