2-Iod-5-nitroanisol

Lieferant: FLUOROCHEM

Synonyme: 1-Iodo-2-methoxy-4-nitrobenzene

149002-25G
FLUO149002-25GEA 0 EUR
FLUO149002-25G
2-Iod-5-nitroanisol
2-Iod-5-nitroanisol
Formel: C₇H₆INO₃
Molecular Weight: 279,03 g/mol
MDL: MFCD00024590
CAS-Nummer: 5458-84-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR