4-Phenoxybenzonitril

Lieferant: FLUOROCHEM

164600-10G
FLUO164600-10GEA 123 EUR
FLUO164600-10G
4-Phenoxybenzonitril
4-Phenoxybenzonitril
Formel: C₁₃H₉NO
MDL: MFCD00017346
CAS-Nummer: 3096-81-9
UN: 3439
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR