2-Chlor-6-iodanilin
Lieferant: FLUOROCHEM
216382-5G
FLUO216382-5GEA
266
EUR
FLUO216382-5G
2-Chlor-6-iodanilin
2-Chlor-6-iodanilin
Formel:
C₆H₅ClIN Molecular Weight: 253,4681 g/mol |
MDL:
MFCD08443948 CAS-Nummer: 84483-28-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...