3-Amino-2-brom-5-chlorpyridin

Lieferant: FLUOROCHEM

Synonyme: 2-Brom-5-chlor-3-pyridylamin

24340-5G
FLUO24340-5GEA 0 EUR
FLUO24340-5G
3-Amino-2-brom-5-chlorpyridin
3-Amino-2-bromo-5-chloropyridine
Formel: C₅H₄BrClN₂
Molecular Weight: 207,46 g/mol
MDL: MFCD07368976
CAS-Nummer: 90902-83-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR