4-(Methylsulfonyl)anilin

Lieferant: FLUOROCHEM

32697-5G
FLUO32697-5GEA 0 EUR
FLUO32697-5G
4-(Methylsulfonyl)anilin
4-(Methylsulfonyl)anilin
Formel: C₇H₉NO₂S
Molecular Weight: 171,21 g/mol
MDL: MFCD00025353
CAS-Nummer: 5470-49-5
EINECS: 226-803-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR