5-Brom-2-furancarbonsäure

Lieferant: FLUOROCHEM

F020339-1G
FLUOF020339-1GEA 31.1 EUR
FLUOF020339-1G
5-Brom-2-furancarbonsäure
5-Bromo-2-furoic acid
Formel: C₅H₃BrO₃
MDL: MFCD00003239
CAS-Nummer: 585-70-6
EINECS: 209-559-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR