4,4-Dimethylpiperidin
Lieferant: FLUOROCHEM
F033146-1G
FLUOF033146-1GEA
188
EUR
FLUOF033146-1G
4,4-Dimethylpiperidin
4,4-Dimethylpiperidin
Formel:
C₇H₁₅N |
MDL:
MFCD04037433 CAS-Nummer: 4045-30-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...