5-Brom-2-fluoranilin

Lieferant: FLUOROCHEM

FL13203.5
FLUOFL13203.5EA 0 EUR
FLUOFL13203.5
5-Brom-2-fluoranilin
5-Brom-2-fluoranilin
Formel: C₆H₅BrFN
Molecular Weight: 190.01 g/mol
MDL: MFCD03094181
CAS-Nummer: 2924-09-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR