2-Amino-4-phenylphenol ≥98,0% (durch GC, titrimetrische Analyse)
Lieferant: FLUOROCHEM
|
Achtung
|
Synonyme:
3-Amino-4-hydroxybiphenyl, 3-Amino-[1,1'-biphenyl]-4-ol
TCA0397.5G
FLUOTCA0397.5GEA
0
EUR
FLUOTCA0397.5G
2-Amino-4-phenylphenol ≥98,0% (durch GC, titrimetrische Analyse)
2-Amino-4-phenylphenol
Formel:
C₁₂H₁₁NO Molecular Weight: 185,2200 g/mol |
MDL:
MFCD00059187 CAS-Nummer: 1134-36-7 EINECS: 214-484-8 |
Spezifikation Testergebnisse
Appearance | White to Gray to Brown powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Solubility in Acetone | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...