CO₂ incubators, Forma™ Steri-Cycle™
Lieferant: THERMO FORMA
381 371TF
THBR381EA 0 EUR
THBR381 FORM371TF
CO₂ incubators, Forma™ Steri-Cycle™
Brutschränke CO₂-Brutschränke
CO₂ incubators with HEPA filtration system to provide continuous protection against airborne contaminants with high-temperature sterilisation cycle that simplifies routine cleaning practices. Made of stainless steel interior with coved corners for easy cleaning, units have stainless steel shelves and supports are designed to support a full product load.
  • Precise temperature control and choice of TC (thermal conductivity) or IR (infrared) sensor
  • Convenient high-temperature decontamination cycle to destroy all contaminants
  • VOC filtration system removes
More Product Information
Neuer Artikel bei VWR!