Crucibles, cylindrical
Lieferant: FRIATEC
201-56030-0400
FRIA201-56030-0400EA 0 EUR
FRIA201-56030-0400
Crucibles, cylindrical
Tiegel
Zirconium oxide, cylindrical.
  • Applied in electrical and mechanical engineering
  • Withstand maximum temperature of 1950 °C
  • Can be used in surface finishing