1-Amino-6-bromisochinolin

Lieferant: FRONTIER SCIENTIFIC

Synonyme: 6-Bromoisoquinolin-1-ylamine

B11656.1G
FRONB11656.1GEA 0 EUR
FRONB11656.1G
1-Amino-6-bromisochinolin
1-Amino-6-bromoisoquinoline
Formel: C₉H₇BrN₂
Molecular Weight: 223.0700 g/mol
MDL: MFCD07374395
CAS-Nummer: 215453-26-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR