Kalium(morpholin-4-ylmethyl)trifluoroborat

Lieferant: FRONTIER SCIENTIFIC

Synonyme: Potassium trifluoro(morpholinomethyl)borate

P10324.5G
FRONP10324.5GEA 0 EUR
FRONP10324.5G
Kalium(morpholin-4-ylmethyl)trifluoroborat
Kalium(morpholin-4-ylmethyl)trifluoroborat
Formel: C₅H₁₀BF₃KNO
Molecular Weight: 207.0435 g/mol
MDL: MFCD10566512
CAS-Nummer: 936329-94-1

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR