2,2,2-Trifluoracetophenon-4-boronsäurepinakolester

Lieferant: FRONTIER SCIENTIFIC

Synonyme: 2,2,2-Trifluoro-1-(4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)ethanone

T12462.1G
FRONT12462.1GEA 0 EUR
FRONT12462.1G
2,2,2-Trifluoracetophenon-4-boronsäurepinakolester
2,2,2-Trifluoracetophenon-4-boronsäurepinakolester
Formel: C₁₄H₁₆BF₃O₃
Molecular Weight: 300.0813 g/mol
MDL: MFCD16628195
CAS-Nummer: 1004294-77-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR