S&T forceps, SuperGrip Tips
Lieferant: FINE SCIENCE TOOLS
00649-11 00632-11
FSCI00649-11EA 437 EUR
FSCI00649-11 FSCI00632-11
S&T forceps, SuperGrip Tips
Pinzetten
SuperGrip tips are processed with MicroCrystals®, a method developed by S&T to infuse tungsten carbide on stainless steel. The formation of this ultra-fine coating guarantees atraumatic handling of tissue while improving the grip dramatically. The gripping surface is similar to a match head.
Recommended when forceps are utilized as needle holders or when slippery tissues might be encountered.