Ring forceps
Lieferant: FINE SCIENCE TOOLS
11103-09 11106-09 11101-09
FSCI11103-09EA 89.9 EUR
FSCI11103-09 FSCI11106-09 FSCI11101-09
Ring forceps
Pinzetten
For transport of tissue and organs. A limit screw prevents excessive applied pressure.