Strabismus scissors, Tungsten Carbide
Lieferant: FINE SCIENCE TOOLS
14574-09 14574-11 14575-09
FSCI14574-09EA 162 EUR
FSCI14574-09 FSCI14574-11 FSCI14575-09
Strabismus scissors, Tungsten Carbide
Scheren
These scissors are made of stainless steel with tungsten carbide inserts.