Vannas spring scissors, 2,5 mm cutting edge
Lieferant: FINE SCIENCE TOOLS
15000-08 15001-08
FSCI15000-08EA 293 EUR
FSCI15000-08 FSCI15001-08
Vannas spring scissors, 2,5 mm cutting edge
Scheren
These stainless steel scissors have been designed to maximise visibility under the microscope by eliminating unnecessary bulk in the blades.
A short taper makes these scissors ideal for small, precise cuts.
Bestellen
TECHNISCHE DATEN
Environmentally Preferable