Ultra fine clipper scissors II forceps style
Lieferant: FINE SCIENCE TOOLS
15300-00
FSCI15300-00EA 381 EUR
FSCI15300-00
Ultra fine clipper scissors II forceps style
Scheren
These stainless steel scissors are best suited for microscopic dissections of soft and mucous tissue.
The minute fineness of the blades allows clear visualisation of the tissue being dissected.