Moria Pascheff-Wolff spring scissors
Lieferant: FINE SCIENCE TOOLS
15371-92
FSCI15371-92EA 728 EUR
FSCI15371-92
Moria Pascheff-Wolff spring scissors
Scheren
These inox scissors have an adjustable screw that allows for varying spring action.