Silver neuro clips
Lieferant: FINE SCIENCE TOOLS
18040-00
FSCI18040-00EA 137 EUR
FSCI18040-00
Silver neuro clips
Erste-Hilfe-Ausstattung Wundklammern
These single-use silver neuro clips are meant for permanent vessel occlusion.
  • Clamp Press: Variable
  • Alloy / Material: Silver
  • Length: 5 mm
  • Not serrated
Bitte beachten All surgical instruments and devices from the Fine Science Tools product range are intended exclusively for use in experimental research laboratories and facilities or in veterinary medicine.