These labels are provided in A4 sheet format for desktop laser printers. They will withstand long-term cryo storage in liquid nitrogen and deep-freeze conditions, as well as sterilisation in steam autoclaves. Wrap-around laser labels also resist abrasion, scratching, as well as spraying and wiping with chemicals and detergents.
- Temperature range: –196 to 150 °C
- Withstands long-term storage in extreme low temperatures (–196 °C)
- Transparent laminating tail provides extra protection against scratching and spraying with chemicals and detergents
- Printable with all major desktop laser printers
- Withstands extreme elevated temperatures (150 °C), able to resist steam autoclaves and boiling water
- Resistant to gamma irradiation (tested up to 50 KGy)
Cryogenic wrap-around laser labels for long-term cryo-storage in liquid nitrogen and deep freeze harsh environments. Temperature range from –196 to 150 °C. Resistant to autoclave, boiling water, as well as spraying with ethanol, IPA, acetone, and other chemicals and detergents. Cryo labels accept desktop laser printing and permanent marker. They are water-resistant, wear and tear proof, will not jam printers, and are resistant to UV and gamma irradiation.