Cryoscope
Lieferant: Gerber Instruments AG
05.25500
GEAG05.25500EA 9100 EUR
GEAG05.25500
Cryoscope
Kryoskope
The dynamic cooling system Cryoscope C1 is ideal for reliable and rapid determination of freezing point in milk acc. to ISO 5764/IDF 108. During the daily inspection of water content in the raw milk, the device works extremely quietly.
  • Easy to calibrate
  • Robust and compact design with touchscreen
  • Adjustable amplitude of the stirrer
  • Time or plateau mode selectable
  • Data output to an external printer and/or PC selectable
  • Worldwide support via internet possible
The dynamic cooling system allows a maximum sample throughput even at ambient temperatures up to 35 °C.
Lieferumfang: System includes standards 408, 512, 600, heat transfer fluid (each 250 ml) and test tubes with ring mark (2,0 ml, 10 pc).